CAS RN 92325-11-6
CAS RN 92325-11-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
5-[2-(Acetyloxy)ethyl]-3-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-4-methyl-2(3H)-thiazolethione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5-[2-(Acetyloxy)ethyl]-3-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-4-methyl-2(3H)-thiazolethione, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.