missing translation for 'onlineSavingsMsg'
Learn More

5-[2-(Acetyloxy)ethyl]-3-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-4-methyl-2(3H)-thiazolethione, TRC

Product Code. 30326200
Change view
Click to view available options
Quantity:
50 mg
250 mg
Unit Size:
250mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30326200 50 mg 50mg
30315035 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30326200 Supplier Toronto Research Chemicals Supplier No. TRCA18713550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-[2-(Acetyloxy)ethyl]-3-[(4-amino-2-methyl-5-pyrimidinyl)methyl]-4-methyl-2(3H)-thiazolethione
CAS 92325-11-6
Molecular Formula C14H18N4O2S2
Quantity 50 mg
Synonym [(4-Amino-2-methyl-5-pyrimidinyl)methyl]-5-(2-hydroxyethyl)-4-methyl-4-thiazoline-2-thione Acetate, 2-(3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-4-methyl-2-thioxo-2,3-dihydrothiazol-5-yl)ethyl acetate
InChI Formula InChI=1S/C14H18N4O2S2/c1-8-12(4-5-20-10(3)19)22-14(21)18(8)7-11-6-16-9(2)17-13(11)15/h6H,4-5,7H2,1-3H3,(H2,15,16,17)
SMILES CC(=O)OCCC1=C(C)N(Cc2cnc(C)nc2N)C(=S)S1
IUPAC Name 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-2-sulfanylidene-1,3-thiazol-5-yl]ethyl acetate
Molecular Weight (g/mol) 338.45
Formula Weight 338.0871
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.