accessibility menu, dialog, popup

UserName

H3CNHOCH3OH

CAS RN 92188-49-3

IUPAC Name: 1-(3-methoxyphenyl)-2-(methylamino)ethan-1-ol
Synonyms: 1-(3-methoxyphenyl)-2-(methylamino)ethanol
Molecular Weight (g/mol): 181.24
Molecular Formula: C10H15NO2
InChi Key: ZIGVURPVSKJORL-UHFFFAOYNA-N
SMILES: CNCC(O)C1=CC(OC)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results