accessibility menu, dialog, popup

UserName

H3CHNONOONOO

CAS RN 92043-81-7

IUPAC Name: N-hexyl-3,5-dinitrobenzamide
Synonyms: N-hexyl-3,5-dinitrobenzamide
Molecular Weight (g/mol): 295.3
Molecular Formula: C13H17N3O5
InChi Key: ULUBCUUBHVXVDB-UHFFFAOYSA-N
SMILES: CCCCCCNC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results