accessibility menu, dialog, popup

UserName

CAS RN 92-69-3

OH

CAS RN 92-69-3

IUPAC Name: [1,1'-biphenyl]-4-ol
Synonyms: phenylphenol P-phenylphenol para-hydroxydiphenyl 4-hydroxydiphenyl p-hydroxydiphenyl paraxenol phenol p-phenyl 4-diphenylol tetrasin P 300 tetrosin P 300
Molecular Weight (g/mol): 170.21
Molecular Formula: C12H10O
InChi Key: YXVFYQXJAXKLAK-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)