missing translation for 'onlineSavingsMsg'
Learn More

p-Hydroxybiphenyl, MP Biomedicals™

Product Code. 11403881 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403881 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11403881 Supplier MP Biomedicals Supplier No. 0215129050

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 92-69-3
Molecular Formula C12H10O
Quantity 50 g
Synonym 4-hydroxybiphenyl, 1,1'-biphenyl-4-ol, biphenyl-4-ol, p-phenylphenol, 4-biphenylol, p-hydroxybiphenyl, 4-hydroxydiphenyl, p-hydroxydiphenyl, p-biphenylol, 4-diphenylol
InChI Key YXVFYQXJAXKLAK-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)O
IUPAC Name 4-phenylphenol
Molecular Weight (g/mol) 170.211
PubChem CID 7103
ChEBI CHEBI:34422
Formula Weight 170.2
Physical Form Solid
Chemical Name or Material p-Hydroxybiphenyl
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.