accessibility menu, dialog, popup

UserName

CAS RN 916420-76-3

IUPAC Name: 3-[2-chloro-3-(trifluoromethyl)phenyl]propanoic acid
Synonyms: 3-[2-chloro-3-(trifluoromethyl)phenyl]propanoic acid
Molecular Weight (g/mol): 252.62
Molecular Formula: C10H8ClF3O2
InChi Key: AYDBHGFWRZTJFK-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=C(Cl)C(=CC=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results