accessibility menu, dialog, popup

UserName

OFFFClOOH

CAS RN 916420-74-1

IUPAC Name: 3-[3-chloro-5-(trifluoromethoxy)phenyl]propanoic acid
Synonyms: 3-[3-chloro-5-(trifluoromethoxy)phenyl]propanoic acid
Molecular Weight (g/mol): 268.62
Molecular Formula: C10H8ClF3O3
InChi Key: OOXKNRUBGDQVIF-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=CC(OC(F)(F)F)=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results