CAS RN 916141-36-1
CAS RN 916141-36-1
IUPAC Name:
N-{[1,1'-biphenyl]-2-yl}-1-{2-[(1-methyl-1H-1,3-benzodiazol-2-yl)sulfanyl]acetyl}pyrrolidine-2-carboxamide
Synonyms:
N-{[1,1'-biphenyl]-2-yl}-1-{2-[(1-methyl-1,3-benzodiazol-2-yl)sulfanyl]acetyl}pyrrolidine-2-carboxamide
Molecular Weight (g/mol):
470.59
Molecular Formula:
C27H26N4O2S
InChi Key:
YSBGRVXJEMSEQY-UHFFFAOYNA-N
SMILES:
CN1C(SCC(=O)N2CCCC2C(=O)NC2=CC=CC=C2C2=CC=CC=C2)=NC2=CC=CC=C12
1
–
2
of
2
results
1
Tocris Bioscience™ TCS 1102
CAS: 916141-36-1 Molecular Formula: C27H26N4O2S
2
TCS 1102, MedChemExpress
MedChemExpress TCS 1102 is a potent, dual orexin receptor antagonist (Ki values are 0.2 and 3 nM for OX2 and OX1 receptors respectively).