accessibility menu, dialog, popup

UserName

CH3NNOBr

CAS RN 915707-60-7

IUPAC Name: 2-(3-bromophenoxy)-6-methylpyrazine
Synonyms: 2-(3-bromophenoxy)-6-methylpyrazine
Molecular Weight (g/mol): 265.11
Molecular Formula: C11H9BrN2O
InChi Key: BCQSIIIFPGJDPS-UHFFFAOYSA-N
SMILES: CC1=CN=CC(OC2=CC(Br)=CC=C2)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results