CAS RN 913835-76-4
CAS RN 913835-76-4
IUPAC Name:
(4-chloro-2-formylphenyl)boronic acid
Synonyms:
4-chloro-2-formylphenylboronic acid
Molecular Weight (g/mol):
184.38
Molecular Formula:
C7H6BClO3
InChi Key:
BTQFQGHQBOWNAJ-UHFFFAOYSA-N
SMILES:
OB(O)C1=C(C=O)C=C(Cl)C=C1
1
–
1
of
1
results
1
4-Chloro-2-formylbenzeneboronic acid, 98%, Thermo Scientific™
CAS: 913835-76-4 Molecular Formula: C7H6BClO3 Molecular Weight (g/mol): 184.382 MDL Number: MFCD08689492 InChI Key: BTQFQGHQBOWNAJ-UHFFFAOYSA-N Synonym: 4-chloro-2-formylphenyl boronic acid,4-chloro-2-formylbenzeneboronic acid,boronicacid, b-4-chloro-2-formylphenyl,2-borono-5-chlorobenzaldehyde,boronic acid, b-4-chloro-2-formylphenyl,acmc-209ra1,2-formyl-4-chlorophenylboronic acid,4-chloro-2-formyl-phenylboronic acid,4-chloro-2-formyl-phenyl boronic acid,4-chloro-2-formylbenzeneboronicacid PubChem CID: 23730425 IUPAC Name: (4-chloro-2-formylphenyl)boronic acid SMILES: B(C1=C(C=C(C=C1)Cl)C=O)(O)O