accessibility menu, dialog, popup

UserName

BFOHNOCH3HOOH

CAS RN 913835-47-9

IUPAC Name: [4-fluoro-3-(methoxycarbamoyl)phenyl]boronic acid
Synonyms: 4-fluoro-3-(methoxycarbamoyl)phenylboronic acid
Molecular Weight (g/mol): 212.97
Molecular Formula: C8H9BFNO4
InChi Key: OJNRHYRWHLOKBF-UHFFFAOYSA-N
SMILES: CONC(=O)C1=C(F)C=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results