accessibility menu, dialog, popup

UserName

CAS RN 912350-00-6

BIFFFK

CAS RN 912350-00-6

IUPAC Name: potassium trifluoro(4-iodophenyl)boranuide
Synonyms: potassium trifluoro(4-iodophenyl)boranuide
Molecular Weight (g/mol): 309.91
Molecular Formula: C6H4BF3IK
InChi Key: WTXNSBOVCAXUGV-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C1=CC=C(I)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results