accessibility menu, dialog, popup

UserName

CAS RN 9041-93-4

IUPAC Name: (3-{[2-(2-{2-[2-(4-{2-[(6-amino-2-{1-[(2-amino-2-carbamoylethyl)amino]-2-carbamoylethyl}-5-methylpyrimidin-4-yl)formamido]-3-{[(2R,3S,4S,5S,6S)-3-{[(2R,3S,4S,5R,6R)-4-(carbamoyloxy)-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-4,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-3-(1H-imidazol-5-yl)propanamido}-3-hydroxy-2-methylpentanamido)-3-hydroxybutanamido]ethyl}-1,3-thiazol-4-yl)-1,3-thiazol-4-yl]formamido}propyl)dimethylsulfanium hydrogen sulfate
Synonyms: bleomycin
Molecular Weight (g/mol): 1512.62
Molecular Formula: C55H85N17O25S4
InChi Key: WUIABRMSWOKTOF-OCBSMOPSSA-N
SMILES: OS([O-])(=O)=O.CC(O)C(NC(=O)C(C)C(O)C(C)NC(=O)C(NC(=O)C1=NC(=NC(N)=C1C)C(CC(N)=O)NCC(N)C(N)=O)C(O[C@@H]1O[C@@H](CO)[C@@H](O)[C@H](O)[C@@H]1O[C@H]1O[C@H](CO)[C@@H](O)[C@H](OC(N)=O)[C@@H]1O)C1=CN=CN1)C(=O)NCCC1=NC(=CS1)C1=NC(=CS1)C(=O)NCCC[S+](C)C
Molecular Weight (g/mol) 
  • (1)
  • (1)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
Color 
  • (1)
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results