accessibility menu, dialog, popup

UserName

CAS RN 90389-34-7

FClNH2HCl

CAS RN 90389-34-7

IUPAC Name: 1-(3-chloro-5-fluorophenyl)methanamine hydrochloride
Synonyms: 1-(3-chloro-5-fluorophenyl)methanamine hydrochloride
Molecular Weight (g/mol): 196.05
Molecular Formula: C7H8Cl2FN
InChi Key: SXKXBIIFCYGVBL-UHFFFAOYSA-N
SMILES: Cl.NCC1=CC(F)=CC(Cl)=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results