CAS RN 9012-36-6
IUPAC Name:
(2S,3R,4S,5R,6R)-2-{[(1S,3S,4S,5S,8R)-3-{[(2S,3R,4S,5S,6R)-2-{[(1S,3R,4S,5S,8R)-3,4-dihydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-3,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy}-4-hydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms:
sepharose 4B C1 4b, sepharose sepharose C1 4B 4b, sepharose C1 agarose gel beads 4b, sepharose
Molecular Weight (g/mol):
630.55
Molecular Formula:
C24H38O19
InChi Key:
MJQHZNBUODTQTK-WKGBVCLCSA-N
SMILES:
OC[C@H]1O[C@@H](O[C@@H]2[C@@H]3CO[C@H]2[C@H](O)[C@H](O[C@H]2[C@@H](O)[C@@H](CO)O[C@@H](O[C@@H]4[C@@H]5CO[C@H]4[C@H](O)[C@H](O)O5)[C@@H]2O)O3)[C@H](O)[C@@H](O)[C@H]1O
1
–
15
of
55
results
1
2
Agarose (Low-Melting, <1kb DNA/RNA/Genetic Analysis Grade), Fisher BioReagents™
Agarose, C12H18O9, CAS Number-9012-36-6, 100g, Gray, Cream, DNase free, 0.15 max., Pass Test, 306.12, 35 deg.C max., 500g/cm2min., Genetic Analysis, Low-Melting, <1kb DNA/RNA, Poly Bottle, Powder/Solid, DNase- and RNase-Free, RT, 0.15% max. | CAS: 9012-36-6 | C12H18O9
3
Agarose (Low-EEO/Multi-Purpose/Molecular Biology Grade), Fisher BioReagents™
CAS: 9012-36-6 Molecular Formula: C12H18O9
4
Agarose (Broad Separation Range for DNA/RNA/Genetic Analysis Grade), Fisher BioReagents™
CAS: 9012-36-6 Molecular Formula: C12H18O9
6
Agarose (Protein Electrophoresis Grade), Fisher BioReagents™
CAS: 9012-36-6 Molecular Formula: C12H18O9
7
Agarose LE, Molecular Biology Grade, Ultrapure
CAS: 9012-36-6 MDL Number: MFCD00081294
8
Thermo Scientific Chemicals Agarose, low EEO
CAS: 9012-36-6 MDL Number: MFCD00081294
9
Thermo Scientific Chemicals Agarose medium EEO, for electrophoresis, (Mr = 0.16 - 0.19)
CAS: 9012-36-6 MDL Number: MFCD00081294
10
11
Thermo Scientific Chemicals Agarose low EEO, for electrophoresis, (Mr <= 0.12)
CAS: 9012-36-6 MDL Number: MFCD00081294
12
Thermo Scientific Chemicals Agarose, Genetic Technology Grade
CAS: 9012-36-6 MDL Number: MFCD00081294
14
Thermo Scientific Chemicals Agarose medium-low EEO, for electrophoresis, (Mr = 0.12)
CAS: 9012-36-6 MDL Number: MFCD00081294
15
Thermo Scientific Chemicals Agarose ME, for electrophoresis of macromolecules
CAS: 9012-36-6 MDL Number: MFCD00081294