accessibility menu, dialog, popup

UserName

CAS RN 9002-91-9

CH3OOOOH3CCH3CH3

CAS RN 9002-91-9

IUPAC Name: 2,4,6,8-tetramethyl-1,3,5,7-tetraoxocane
Synonyms: metaldehyde metaldehyde (tetramer)
Molecular Weight (g/mol): 176.21
Molecular Formula: C8H16O4
InChi Key: GKKDCARASOJPNG-UHFFFAOYSA-N
SMILES: CC1OC(C)OC(C)OC(C)O1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
Concentration 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results