Learn More
Metaldehyde in Acetonitrile 100μg/mL, Fisher Chemical™
BIOPHARMA
Product Code. 12949903 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12949903 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12949903 Supplier Fisher Chemical Supplier No. P831350
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Packaging Ampoule(s)
CAS 9002-91-9
Molecular Formula C8H16O4
Concentration 100 μg/mL
UN Number 1648
InChI Key GKKDCARASOJPNG-UHFFFAOYSA-N
SMILES CC1OC(OC(OC(O1)C)C)C
IUPAC Name 2,4,6,8-tetramethyl-1,3,5,7-tetraoxocane
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 176.212
PubChem CID 61021
Quantity 1 mL
ChEBI CHEBI:81931
Chemical Name or Material Metaldehyde
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Standard solution Metaldehyde 100ug/ml in acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.