CAS RN 900019-38-7
IUPAC Name:
methyl 6-chloroimidazo[1,2-a]pyridine-3-carboxylate
Synonyms:
methyl 6-chloroimidazo[1,2-a]pyridine-3-carboxylate
Molecular Weight (g/mol):
210.62
Molecular Formula:
C9H7ClN2O2
InChi Key:
UFRZEIOOOFHTPS-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CN=C2C=CC(Cl)=CN12
1
–
1
of
1
results
1
Methyl 6-chloroimidazo[1,2-a]pyridine-3-carboxylate, 95%, Thermo Scientific™
CAS: 900019-38-7 Molecular Formula: C9H7ClN2O2 Molecular Weight (g/mol): 210.617 MDL Number: MFCD07021508 InChI Key: UFRZEIOOOFHTPS-UHFFFAOYSA-N Synonym: methyl 6-chloroimidazo 1,2-a pyridine-3-carboxylate,methyl 6-chloroimidazo-1,2-a pyridine-3-carboxylate,6-chloroimidazo 1,2-a pyridine-3-carboxylic acid methyl ester,imidazo 1,2-a pyridine-3-carboxylic acid,6-chloro-,methyl ester PubChem CID: 16413638 IUPAC Name: methyl 6-chloroimidazo[1,2-a]pyridine-3-carboxylate SMILES: COC(=O)C1=CN=C2N1C=C(C=C2)Cl