accessibility menu, dialog, popup

UserName

CAS RN 89379-79-3

H3CONCl

CAS RN 89379-79-3

IUPAC Name: 3-chloro-4-methoxypyridine
Synonyms: 3-chloro-4-methoxypyridine
Molecular Weight (g/mol): 143.57
Molecular Formula: C6H6ClNO
InChi Key: OFJIIJJCVDJQFW-UHFFFAOYSA-N
SMILES: COC1=C(Cl)C=NC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results