accessibility menu, dialog, popup

UserName

CH3NH2FF

CAS RN 886503-79-3

IUPAC Name: 2,3-difluoro-4-methylaniline
Synonyms: 2,3-difluoro-4-methylaniline
Molecular Weight (g/mol): 143.14
Molecular Formula: C7H7F2N
InChi Key: HMDHQYKNBZEFTH-UHFFFAOYSA-N
SMILES: CC1=CC=C(N)C(F)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results