CAS RN 886498-07-3
IUPAC Name:
3-bromo-5-(trifluoromethoxy)benzaldehyde
Synonyms:
3-bromo-5-(trifluoromethoxy)benzaldehyde
Molecular Weight (g/mol):
269.02
Molecular Formula:
C8H4BrF3O2
InChi Key:
ALIVUZVZCLVJQN-UHFFFAOYSA-N
SMILES:
FC(F)(F)OC1=CC(Br)=CC(C=O)=C1
1
–
1
of
1
results
1
3-Bromo-5-(trifluoromethoxy)benzaldehyde, 97%, Thermo Scientific™
CAS: 886498-07-3 Molecular Formula: C8H4BrF3O2 Molecular Weight (g/mol): 269.017 MDL Number: MFCD04116040 InChI Key: ALIVUZVZCLVJQN-UHFFFAOYSA-N Synonym: 3-bromo-5-trifluoromethoxy benzaldehyde,acmc-209qvy,3-bromo-5-trifluoromethoxy-benzaldehyde PubChem CID: 5006711 IUPAC Name: 3-bromo-5-(trifluoromethoxy)benzaldehyde SMILES: C1=C(C=C(C=C1OC(F)(F)F)Br)C=O