accessibility menu, dialog, popup

UserName

CAS RN 88321-09-9

CH3OO(S)O(S)OHN(S)CH3H3COHNCH3CH3

CAS RN 88321-09-9

IUPAC Name: ethyl (2S,3S)-3-{[(1S)-3-methyl-1-[(3-methylbutyl)carbamoyl]butyl]carbamoyl}oxirane-2-carboxylate
Synonyms: ethyl (2S,3S)-3-{[(1S)-3-methyl-1-[(3-methylbutyl)carbamoyl]butyl]carbamoyl}oxirane-2-carboxylate
Molecular Weight (g/mol): 342.44
Molecular Formula: C17H30N2O5
InChi Key: SRVFFFJZQVENJC-IHRRRGAJSA-N
SMILES: CCOC(=O)[C@H]1O[C@@H]1C(=O)N[C@@H](CC(C)C)C(=O)NCCC(C)C
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Color 
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results