accessibility menu, dialog, popup

UserName

CAS RN 882679-10-9

IUPAC Name: 2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
Synonyms: 2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
Molecular Weight (g/mol): 266.08
Molecular Formula: C13H16BFO4
InChi Key: DXOXOBZCGBHKJS-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(C(O)=O)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results