accessibility menu, dialog, popup

UserName

OHOClClNOO

CAS RN 88-86-8

IUPAC Name: 2,5-dichloro-3-nitrobenzoic acid
Synonyms: dinoben
Molecular Weight (g/mol): 236
Molecular Formula: C7H3Cl2NO4
InChi Key: AUAXYMOBWXOEQD-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(Cl)=CC(=C1Cl)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results