accessibility menu, dialog, popup

UserName

CAS RN 875770-34-6

SOHNOOONNNNHOHN

CAS RN 875770-34-6

IUPAC Name: N-(2-{2-[2-(2-azidoethoxy)ethoxy]ethoxy}ethyl)-5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanamide
Synonyms: N-(2-{2-[2-(2-azidoethoxy)ethoxy]ethoxy}ethyl)-5-{2-oxo-hexahydrothieno[3,4-d]imidazol-4-yl}pentanamide
Molecular Weight (g/mol): 444.55
Molecular Formula: C18H32N6O5S
InChi Key: ZWFOOMQCYIGZBE-UHFFFAOYNA-N
SMILES: [N-]=[N+]=NCCOCCOCCOCCNC(=O)CCCCC1SCC2NC(=O)NC12
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results