accessibility menu, dialog, popup

UserName

BOHNH3CCH3FHOOH

CAS RN 874289-49-3

IUPAC Name: {2-fluoro-5-[(propan-2-yl)carbamoyl]phenyl}boronic acid
Synonyms: 2-fluoro-5-(isopropylcarbamoyl)phenylboronic acid
Molecular Weight (g/mol): 225.03
Molecular Formula: C10H13BFNO3
InChi Key: NCRUGKRIPMSQHI-UHFFFAOYSA-N
SMILES: CC(C)NC(=O)C1=CC(B(O)O)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results