accessibility menu, dialog, popup

UserName

BClOOCH3HOOH

CAS RN 871329-55-4

IUPAC Name: [5-chloro-2-(ethoxycarbonyl)phenyl]boronic acid
Synonyms: 5-chloro-2-(ethoxycarbonyl)phenylboronic acid
Molecular Weight (g/mol): 228.44
Molecular Formula: C9H10BClO4
InChi Key: SGPMCOBGNHZGJJ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C=C(Cl)C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results