accessibility menu, dialog, popup

UserName

BOOCH3CH3CH3CH3HN

CAS RN 871125-67-6

IUPAC Name: 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
Synonyms: 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
Molecular Weight (g/mol): 293.17
Molecular Formula: C18H20BNO2
InChi Key: RLSJGSFDSSYNPL-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC2=C(C=C1)C1=C(N2)C=CC=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results