accessibility menu, dialog, popup

UserName

CAS RN 870704-16-8

IUPAC Name: 1-(2,3-dichloro-6-fluorophenyl)ethan-1-one
Synonyms: 1-(2,3-dichloro-6-fluorophenyl)ethanone
Molecular Weight (g/mol): 207.03
Molecular Formula: C8H5Cl2FO
InChi Key: VHFIYTUVCUSGMN-UHFFFAOYSA-N
SMILES: CC(=O)C1=C(F)C=CC(Cl)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results