CAS RN 87-51-4
1H-Indole-3-acetic acid, 98%
CAS: 87-51-4 Molecular Formula: C10H9NO2 Molecular Weight (g/mol): 175.19 MDL Number: MFCD00005636 InChI Key: SEOVTRFCIGRIMH-UHFFFAOYSA-N Synonym: indole-3-acetic acid,3-indoleacetic acid,heteroauxin,indoleacetic acid,1h-indole-3-acetic acid,2-1h-indol-3-yl acetic acid,rhizopin,1h-indol-3-ylacetic acid,indol-3-ylacetic acid,rhizopon a PubChem CID: 802 ChEBI: CHEBI:16411 IUPAC Name: 2-(1H-indol-3-yl)acetic acid SMILES: OC(=O)CC1=CNC2=CC=CC=C12
Indole-3-acetic acid, 98+%
CAS: 87-51-4 Molecular Formula: C10H9NO2 Molecular Weight (g/mol): 175.19 MDL Number: MFCD00005636 InChI Key: SEOVTRFCIGRIMH-UHFFFAOYSA-N Synonym: indole-3-acetic acid,3-indoleacetic acid,heteroauxin,indoleacetic acid,1h-indole-3-acetic acid,2-1h-indol-3-yl acetic acid,rhizopin,1h-indol-3-ylacetic acid,indol-3-ylacetic acid,rhizopon a PubChem CID: 802 ChEBI: CHEBI:16411 IUPAC Name: 2-(1H-indol-3-yl)acetic acid SMILES: OC(=O)CC1=CNC2=CC=CC=C12
Indole-3-acetic acid, 99.2%, MP Biomedicals™
CAS: 87-51-4 Molecular Formula: C10H9NO2 Molecular Weight (g/mol): 175.19 MDL Number: MFCD00005636 InChI Key: SEOVTRFCIGRIMH-UHFFFAOYSA-N Synonym: indole-3-acetic acid,3-indoleacetic acid,heteroauxin,indoleacetic acid,1h-indole-3-acetic acid,2-1h-indol-3-yl acetic acid,rhizopin,1h-indol-3-ylacetic acid,indol-3-ylacetic acid,rhizopon a PubChem CID: 802 ChEBI: CHEBI:16411 IUPAC Name: 2-(1H-indol-3-yl)acetic acid SMILES: OC(=O)CC1=CNC2=CC=CC=C12
Indole-3-acetic Acid, Millipore™
CAS: 87-51-4
Indoleacetic Acid, TRC
CAS: 87-51-4 Molecular Formula: C10 H9 N O2 Molecular Weight (g/mol): 175.18 Synonym: 1H-Indole-3-acetic acid,Indole-3-acetic acid (8CI),(1H-Indol-3-yl)acetic acid,2-(1H-Indol-3-yl)acetic acid,2-(3-Indolyl)acetic acid,3-(Carboxymethyl)-1H-indole,3-(Carboxymethyl)indole,3-IAA,3-Indolylmethylcarboxylic acid,Bioenraiz,GAP,Heteroauxin,IAA,Indoleacetic acid,NSC 3787,Noclosan,Rhizopin,α-IAA,β-IAA,β-Indoleacetic acid,3-Indolylacetic acid,3-Indolyl acetic acid IUPAC Name: 2-(1H-indol-3-yl)acetic acid SMILES: OC(=O)Cc1c[nH]c2ccccc12
R&D Systems™ Auxin
A Tocris product, part of R&D Systems. Chemical dimerizer used in auxin-inducible degron (AID) system; phytohormone
3-Indolyl acetic acid 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
MedChemExpress 3-Indoleacetic acid
MedChemExpress 3-Indoleacetic acid (Indole-3-acetic acid) is the most common natural plant growth hormone of the auxin class. It can be added to cell culture medium to induce plant cell elongation and division.