accessibility menu, dialog, popup

UserName

H3CNCH3

CAS RN 86-56-6

IUPAC Name: N,N-dimethylnaphthalen-1-amine
Synonyms: N,N-dimethyl-1-naphthylamine
Molecular Weight (g/mol): 171.24
Molecular Formula: C12H13N
InChi Key: AJUXDFHPVZQOGF-UHFFFAOYSA-N
SMILES: CN(C)C1=C2C=CC=CC2=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (2)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)
  • (2)
  • (6)