missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-α-naphthylamine, MP Biomedicals

Product Code. 11473125 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473125 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11473125 Supplier MP Biomedicals Supplier No. 0521375505

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 86-56-6
Molecular Formula C12H13N
Quantity 5 g
Synonym n,n-dimethyl-1-naphthylamine, 1-dimethylaminonaphthalene, 1-naphthalenamine, n,n-dimethyl, n,n-dimethyl-1-naphthalenamine, n,n-dimethyl-1-napthylamine, dimethyl 1-naphthyl amine, alpha-dimethylaminonaphthalene, 1-naphthylamine, n,n-dimethyl, dimethyl-alpha-naphthylamine, n,n-dimethyl-1-naftylamin
InChI Key AJUXDFHPVZQOGF-UHFFFAOYSA-N
SMILES CN(C)C1=CC=CC2=CC=CC=C21
IUPAC Name N,N-dimethylnaphthalen-1-amine
Molecular Weight (g/mol) 171.243
PubChem CID 6848
Formula Weight 171.24
Chemical Name or Material N, N-Dimethyl-α-naphthylamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.