accessibility menu, dialog, popup

UserName

CAS RN 856866-72-3

CH3NNNNNNOOOHF

CAS RN 856866-72-3

IUPAC Name: 3-{3-fluoro-4-[6-(2-methyl-2H-1,2,3,4-tetrazol-5-yl)pyridin-3-yl]phenyl}-5-(hydroxymethyl)-1,3-oxazolidin-2-one
Synonyms: 3-{3-fluoro-4-[6-(2-methyl-1,2,3,4-tetrazol-5-yl)pyridin-3-yl]phenyl}-5-(hydroxymethyl)-1,3-oxazolidin-2-one
Molecular Weight (g/mol): 370.34
Molecular Formula: C17H15FN6O3
InChi Key: XFALPSLJIHVRKE-UHFFFAOYNA-N
SMILES: CN1N=NC(=N1)C1=NC=C(C=C1)C1=C(F)C=C(C=C1)N1CC(CO)OC1=O
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results