accessibility menu, dialog, popup

UserName

H3COBrFFF

CAS RN 853367-87-0

IUPAC Name: 4-(bromomethyl)-2-methoxy-1-(trifluoromethyl)benzene
Synonyms: 4-(bromomethyl)-2-methoxy-1-(trifluoromethyl)benzene
Molecular Weight (g/mol): 269.06
Molecular Formula: C9H8BrF3O
InChi Key: MUVUCILNTIWNQL-UHFFFAOYSA-N
SMILES: COC1=C(C=CC(CBr)=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results