accessibility menu, dialog, popup

UserName

H3CONCH3(S)OH(R)OH(R)OH(R)OHOH

CAS RN 85316-98-9

IUPAC Name: N-methyl-N-[(2S,3R,4R,5R)-2,3,4,5,6-pentahydroxyhexyl]octanamide
Synonyms: N-methyl-N-[(2S,3R,4R,5R)-2,3,4,5,6-pentahydroxyhexyl]octanamide
Molecular Weight (g/mol): 321.41
Molecular Formula: C15H31NO6
InChi Key: SBWGZAXBCCNRTM-CTHBEMJXSA-N
SMILES: CCCCCCCC(=O)N(C)C[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results