accessibility menu, dialog, popup

UserName

CAS RN 850608-87-6

OFFFSNH2NHCl

CAS RN 850608-87-6

IUPAC Name: 6-(trifluoromethoxy)-1,3-benzothiazol-2-amine hydrochloride
Synonyms: riluzole hydrochloride
Molecular Weight (g/mol): 270.65
Molecular Formula: C8H6ClF3N2OS
InChi Key: QEAOELIJQRYJJS-UHFFFAOYSA-N
SMILES: Cl.NC1=NC2=CC=C(OC(F)(F)F)C=C2S1
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results