CAS RN 84964-24-9
CAS RN 84964-24-9
IUPAC Name:
1-(3-bromophenyl)piperidine
Synonyms:
1-(3-bromophenyl)piperidine
Molecular Weight (g/mol):
240.14
Molecular Formula:
C11H14BrN
InChi Key:
PMTXPOLLPCSCBE-UHFFFAOYSA-N
SMILES:
BrC1=CC=CC(=C1)N1CCCCC1