accessibility menu, dialog, popup

UserName

CAS RN 845894-03-3

CH3H3CCH3OONOF

CAS RN 845894-03-3

IUPAC Name: tert-butyl 3-fluoro-4-oxopyrrolidine-1-carboxylate
Synonyms: tert-butyl 3-fluoro-4-oxopyrrolidine-1-carboxylate
Molecular Weight (g/mol): 203.21
Molecular Formula: C9H14FNO3
InChi Key: HPHVNLXMQXEDFX-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CC(F)C(=O)C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results