CAS RN 842-18-2
CAS RN 842-18-2
IUPAC Name:
dipotassium 7-hydroxynaphthalene-1,3-disulfonate
Synonyms:
dipotassium 7-hydroxynaphthalene-1,3-disulfonate
Molecular Weight (g/mol):
380.47
Molecular Formula:
C10H6K2O7S2
InChi Key:
LKDMVWRBMGEEGR-UHFFFAOYSA-L
SMILES:
[K+].[K+].OC1=CC=C2C=C(C=C(C2=C1)S([O-])(=O)=O)S([O-])(=O)=O
1
–
2
of
2
results
1
Dipotassium 7-Hydroxy-1,3-naphthalenedisulfonate, Mikromol
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
7-Hydroxy-1,3-naphthalenedisulfonic Acid Dipotassium Salt, TRC
CAS: 842-18-2 Molecular Formula: C10 H6 O7 S2 . 2 K Molecular Weight (g/mol): 380.48 Synonym: 7-Hydroxy-1,3-Naphthalenedisulfonic acid potassium salt (1:2),1,3-Naphthalenedisulfonic acid, 7-hydroxy-, dipotassium salt (8CI,9CI),2-Naphthol-6,8-disulfonic acid dipotassium salt,Dipotassium 2-naphthol-6,8-disulfonate IUPAC Name: dipotassium;7-hydroxynaphthalene-1,3-disulfonate SMILES: [K+].[K+].Oc1ccc2cc(cc(c2c1)S(=O)(=O)[O-])S(=O)(=O)[O-]