missing translation for 'onlineSavingsMsg'
Learn More

Dipotassium 7-Hydroxy-1,3-naphthalenedisulfonate, Mikromol

Product Code. 30461052
Change view
Click to view available options
Quantity:
25 mg
4 x 25 mg
Unit Size:
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30461052 4 x 25 mg -
30051407 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30461052 Supplier Mikromol Supplier No. MM3705.0100254AMP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Dipotassium 7-Hydroxy-1,3-naphthalenedisulfonate, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 7-Hydroxy-1,3-naphthalenedisulfonic Acid Potassium Salt
CAS 842-18-2
Analyte or Component Names 7-Hydroxy-1,3-naphthalenedisulfonic Acid Potassium Salt
Shipping Condition Room Temperature
Molecular Formula C10 H6 O7 S2 . 2 K
Certifications/Compliance ISO 9001
Synonym 7-Hydroxy-1,3-Naphthalenedisulfonic acid potassium salt (1:2), 1,3-Naphthalenedisulfonic acid, 7-hydroxy-, dipotassium salt (8CI,9CI), 2-Naphthol-6,8-disulfonic acid dipotassium salt, Dipotassium 2-naphthol-6,8-disulfonate
InChI Formula InChI=1S/C10H8O7S2.2K/c11-7-2-1-6-3-8(18(12,13)14)5-10(9(6)4-7)19(15,16)17;;/h1-5,11H,(H,12,13,14)(H,15,16,17);;/q;2*+1/p-2
Solution Type Neat
SMILES [K+].[K+].Oc1ccc2cc(cc(c2c1)S(=O)(=O)[O-])S(=O)(=O)[O-]
IUPAC Name dipotassium;7-hydroxynaphthalene-1,3-disulfonate
Molecular Weight (g/mol) 380.48
Quantity 4 x 25 mg
Formula Weight 379.8829 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.