accessibility menu, dialog, popup

UserName

CAS RN 83654-05-1

ONHHNOONON

CAS RN 83654-05-1

IUPAC Name: cyclohexylideneamino N-[6-({[(cyclohexylideneamino)oxy]carbonyl}amino)hexyl]carbamate
Synonyms: cyclohexylideneamino N-[6-({[(cyclohexylideneamino)oxy]carbonyl}amino)hexyl]carbamate
Molecular Weight (g/mol): 394.52
Molecular Formula: C20H34N4O4
InChi Key: RXSVYGIGWRDVQC-UHFFFAOYSA-N
SMILES: O=C(NCCCCCCNC(=O)ON=C1CCCCC1)ON=C1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results