CAS RN 827-94-1
IUPAC Name:
2,6-dibromo-4-nitroaniline
Synonyms:
2,6-dibromo-4-nitroaniline
Molecular Weight (g/mol):
295.92
Molecular Formula:
C6H4Br2N2O2
InChi Key:
YMZIFDLWYUSZCC-UHFFFAOYSA-N
SMILES:
NC1=C(Br)C=C(C=C1Br)[N+]([O-])=O
1
–
4
of
4
results
1
2,6-Dibromo-4-nitroaniline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
2,6-Dibromo-4-nitroaniline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
2,6-Dibromo-4-nitroaniline, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.