missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2,6-Dibromo-4-nitroaniline, 97+%

Product Code. 11477947
Change view
Click to view available options
Quantity:
250g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11477947 250g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11477947 Supplier Alfa Aesar™ Supplier No. A16603.30
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 827-94-1
Molecular Formula C6H4Br2N2O2
MDL Number MFCD00007639
Quantity 250g
Beilstein 2110915
Synonym benzenamine, 2,6-dibromo-4-nitro, aniline, 2,6-dibromo-4-nitro, unii-gwi915pon9, gwi915pon9, 2,6-dibromo-4-nitro-aniline, 2,6-dibromo-4-nitrophenylamine, 2,6-dibromo-4-nitro-phenylamine, pubchem2330, dsstox_cid_31146, dsstox_gsid_52573
InChI Key YMZIFDLWYUSZCC-UHFFFAOYSA-N
SMILES NC1=C(Br)C=C(C=C1Br)[N+]([O-])=O
IUPAC Name 2,6-dibromo-4-nitroaniline
Molecular Weight (g/mol) 295.92
PubChem CID 13231
Formula Weight 295.92
Percent Purity ≥97%
Chemical Name or Material 2,6-Dibromo-4-nitroaniline
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.