accessibility menu, dialog, popup

UserName

CAS RN 824395-67-7

PP

CAS RN 824395-67-7

IUPAC Name: 1-[2-(2,5-diphenylphospholan-1-yl)ethyl]-2,5-diphenylphospholane
Synonyms: 1-[2-(2,5-diphenylphospholan-1-yl)ethyl]-2,5-diphenylphospholane
Molecular Weight (g/mol): 506.61
Molecular Formula: C34H36P2
InChi Key: VHHAZLMVLLIMHT-UHFFFAOYNA-N
SMILES: C(CP1C(CCC1C1=CC=CC=C1)C1=CC=CC=C1)P1C(CCC1C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results