Learn More
(+)-1,2-Bis((2S,5S)-2,5-diphenylphospholano)ethane, TRC
Specifications
Specifications
| Chemical Name or Material | (+)-1,2-Bis((2S,5S)-2,5-diphenylphospholano)ethane |
| CAS | 824395-67-7 |
| Molecular Formula | C34H36P2 |
| Quantity | 100 mg |
| Synonym | (2S,2'S,5S,5'S)-1,1'-(1,2-Ethanediyl)bis[2,5-diphenylphospholane], (S,S)-Ph-BPE |
| InChI Formula | InChI=1S/C34H36P2/c1-5-13-27(14-6-1)31-21-22-32(28-15-7-2-8-16-28)35(31)25-26-36-33(29-17-9-3-10-18-29)23-24-34(36)30-19-11-4-12-20-30/h1-20,31-34H,21-26H2/t31-,32-,33-,34-/m0/s1 |
| SMILES | C(CP1[C@@H](CC[C@H]1c2ccccc2)c3ccccc3)P4[C@@H](CC[C@H]4c5ccccc5)c6ccccc6 |
| IUPAC Name | (2S,5S)-1-[2-[(2S,5S)-2,5-diphenylphospholan-1-yl]ethyl]-2,5-diphenylphospholane |
| Molecular Weight (g/mol) | 506.6 |
| Formula Weight | 506.2292 |