accessibility menu, dialog, popup

UserName

SnOOOO

CAS RN 814-94-8

IUPAC Name: λ²-tin(2+) oxalate
Synonyms: λ²-tin(2+) oxalate
Molecular Weight (g/mol): 206.73
Molecular Formula: C2O4Sn
InChi Key: OQBLGYCUQGDOOR-UHFFFAOYSA-L
SMILES: [Sn++].[O-]C(=O)C([O-])=O
Molecular Weight (g/mol) 
  • (5)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results