Learn More
Tin(II) Oxalate, TRC
Specifications
Specifications
| CAS | 814-94-8 |
| Quantity | 10 g |
| Molecular Formula | C2O4Sn |
| Synonym | Ethanedioic Acid Tin(2+) Salt, Oxalic Acid Tin(2+) Salt, Tin Oxalate (SnC2O4), Fascat 2001, Stannous Oxalate, Tin(2+) Oxalate |
| InChI Formula | InChI=1S/C2H2O4.Sn/c3-1(4)2(5)6;/h(H,3,4)(H,5,6);/q;+2/p-2 |
| SMILES | O=C1O[Sn]OC1=O |
| IUPAC Name | 1,3,2λ^{2}-dioxastannolane-4,5-dione |
| Molecular Weight (g/mol) | 206.73 |
| Formula Weight | 207.8819 |
| Chemical Name or Material | Tin(II) Oxalate |