CAS RN 81177-92-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(1-Ethoxyvinyl)trimethylsilane, TRC
CAS: 81177-92-6 Molecular Formula: C7H16OSi Molecular Weight (g/mol): 144.29 IUPAC Name: 1-ethoxyethenyl(trimethyl)silane SMILES: CCOC(=C)[Si](C)(C)C