accessibility menu, dialog, popup

UserName

H3COONH2n

CAS RN 80506-64-5

IUPAC Name: 2,5,8,11,14,17,20,23,26,29,32,35-dodecaoxaheptatriacontan-37-amine
Synonyms: 2,5,8,11,14,17,20,23,26,29,32,35-dodecaoxaheptatriacontan-37-amine
Molecular Weight (g/mol): 559.69
Molecular Formula: C25H53NO12
InChi Key: MSKSQCLPULZWNO-UHFFFAOYSA-N
SMILES: COCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCN
Quantity 
  • (3)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results